Please login to your online account to display your discounted pricing

anti-Rad1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acid 133-148 of human Rad1 (accession number O60671)
Antigen Rad1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056320

 
novus biologicals canada
NB10056320
Unlabeled 0.1mg Each for $255.80

NB10056320T

 
novus biologicals canada
NB10056320SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acid 133-148 of human Rad1 (accession number O60671)
Antigen Rad1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. O60671
Gene ID (Entrez) 5810
Formulation PBS
Concentration 0.5mg/mL