Please login to your online account to display your discounted pricing

anti-Rabbit Prion protein, Clone: C16-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171679SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171679SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Sheep
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rabbit
Clone C16-S
Immunogen Peptide derived the C-terminus of third a-helical domain of human prion protein.
Antigen Prion protein
Concentration 1.2mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 5621
Formulation 20mM Tris-HCl, pH 8.0. 10mg/mL BSA.
Gene Accession No. P04156
Monoclonal or Polyclonal Monoclonal
Quantity 0.02mL
Regulatory Status RUO
Format Affinity Purified