Please login to your online account to display your discounted pricing

anti-Rabbit Prion protein, Clone: C16-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171679

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171679

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Immunogen Peptide derived the C-terminus of third a-helical domain of human prion protein.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Sheep
Antigen Prion protein
Formulation 20mM Tris-HCl, pH 8.0. 10mg/mL BSA.
Gene Accession No. P04156
Primary or Secondary Primary
Gene ID (Entrez) 5621
Concentration 1.2mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone C16-S
Format Affinity Purified