Please login to your online account to display your discounted pricing

anti-Rabbit Prion protein, Clone: C16-S, Novus Biologicals™

$214.00 - $719.00

Specifications

Clone C16-S
Host Species Rabbit
Immunogen Peptide derived the C-terminus of third a-helical domain of human prion protein.
Antigen Prion protein
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171679

 
novus biologicals canada
NBP171679
Unlabeled 0.1mL Each for $719.00

NBP171679SS

 
novus biologicals canada
NBP171679SS
Unlabeled 0.02mL Each for $214.00
Description & Specifications

Specifications

Clone C16-S
Host Species Rabbit
Immunogen Peptide derived the C-terminus of third a-helical domain of human prion protein.
Antigen Prion protein
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P04156
Gene ID (Entrez) 5621
Formulation 20mM Tris-HCl, pH 8.0. 10mg/mL BSA.
Concentration 1.2mg/mL