Please login to your online account to display your discounted pricing

anti-Rabbit F4/80/EMR1, Clone: SP115, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2125061ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2125061M

  • / Each
 

Description & Specifications

Specifications

Antigen F4/80/EMR1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP115
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 1% BSA, pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Isotype IgG
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 13733
Test Specificity Predicted to cross react with Rat.
Target Species Human
Target Species Murine