Please login to your online account to display your discounted pricing

anti-Rabbit F4/80/EMR1, Clone: SP115, Novus Biologicals™

$139.00 - $758.71

Specifications

Clone SP115
Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Antigen F4/80/EMR1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP212506SS

 
novus biologicals canada
NBP212506SS
Unlabeled 0.02mL Each for $139.00

NBP2125061M

 
novus biologicals canada
NBP2125061ML
Unlabeled 1mL Each for $758.71
Description & Specifications

Specifications

Clone SP115
Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Antigen F4/80/EMR1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Predicted to cross react with Rat.
Gene ID (Entrez) 13733
Formulation PBS with 1% BSA, pH 7.6