Please login to your online account to display your discounted pricing

anti-Rabbit F4/80/EMR1, Clone: SP115, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP212506SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP212506SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen F4/80/EMR1
Quantity 0.02mL
Host Species Rabbit
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Rat.
Conjugate Unlabeled
Gene ID (Entrez) 13733
Primary or Secondary Primary
Formulation PBS with 1% BSA, pH 7.6
Immunogen Synthetic peptide derived from N-terminus of mouse F4/80 protein.
Format Purified
Clone SP115