Please login to your online account to display your discounted pricing

anti-Rabbit Cytokeratin 5, Clone: EP1601Y, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11056916SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11056916T

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 5
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Clone EP1601Y
Concentration 0.004mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Supernatant
Formulation 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, and 0.05% BSA.
Gene Accession No. P13647
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues on the C-terminus of human CK-5 was used as an immunogen.
Isotype IgG
Quantity 0.02mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3852
Test Specificity Mouse cross reactivity tested by western blot and Immunohistochemistry. Rat cross reactivity tested by Western Blot only.