Please login to your online account to display your discounted pricing

anti-Rabbit Cytokeratin 5, Clone: EP1601Y, Novus Biologicals™

$117.00 - $321.00

Specifications

Clone EP1601Y
Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to residues on the C-terminus of human CK-5 was used as an immunogen.
Antigen Cytokeratin 5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11056916T

 
novus biologicals canada
NB11056916SS
Unlabeled 0.02mL Each for $117.00

NB11056916

 
novus biologicals canada
NB11056916
Unlabeled 0.1mL Each for $321.00
Description & Specifications

Specifications

Clone EP1601Y
Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to residues on the C-terminus of human CK-5 was used as an immunogen.
Antigen Cytokeratin 5
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity Mouse cross reactivity tested by western blot and Immunohistochemistry. Rat cross reactivity tested by Western Blot only.
Gene Accession No. P13647
Gene ID (Entrez) 3852
Formulation 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, and 0.05% BSA.
Concentration 0.004mg/mL