Please login to your online account to display your discounted pricing

anti-Rab7a, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105048

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP105048

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P51149
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7879
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Sheep
Primary or Secondary Primary
Antigen Rab7a
Host Species Rabbit
Format Antiserum
Immunogen Recombinant full length human RAB7A protein [Swiss-Prot No. P51149]