Please login to your online account to display your discounted pricing

anti-Rab7a, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Recombinant full length human RAB7A protein [Swiss-Prot No. P51149]
Antigen Rab7a
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP105048

 
novus biologicals canada
NBP105048
Unlabeled 0.1mL Each for $329.00

NBP105048SS

 
novus biologicals canada
NBP105048SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Recombinant full length human RAB7A protein [Swiss-Prot No. P51149]
Antigen Rab7a
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. P51149
Gene ID (Entrez) 7879