Please login to your online account to display your discounted pricing

anti-Rab7a, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105048SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP105048SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. P51149
Gene ID (Entrez) 7879
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Sheep
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Antigen Rab7a
Quantity 0.025mL
Host Species Rabbit
Immunogen Recombinant full length human RAB7A protein [Swiss-Prot No. P51149]