Please login to your online account to display your discounted pricing

anti-Pyruvate Carboxylase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149536

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149536

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5091
Formulation PBS, 0.1% BSA, 50% glycerol
Gene Accession No. P11498
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.22mg/mL
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Pyruvate Carboxylase
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A genomic peptide made to an internal region of the human Pyruvate Carboxylase protein (within residues 930-1050). [Swiss-Prot P11498]
Test Specificity Human and mouse. Immunogen has 96% identity to bovine and porcine. Rat reactivity reported in scientific literature (PMID: 24333689).
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit