missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PWP1 homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PWP1 protein.
Supplier: Abnova Corporation H00011137B01
Description
Sequence: MNRSRQVTCVAWVRCGVAKETPDKVELSKEEVKRLIAEAKEKLQEEGGGSDEEETGSPSEDGMQSARTQARPREPLEDGDPEDDRTLDDDELAEYDLDKYDEEGDPDAETLGESLLGLTVYGSNDQDPYVTLKDTEQYEREDFLIKPSDNLIVCGRAEQDQCNLDVHVYNQEEDSFYVHHDILLSAYPLSVEWLNFDPSPDDSTGNYIAVGNMTPVIEVWDLDIVDSLEPVFTLGSKLSKKKKKKGKKSSSAEGHTDAVLDLSWNKLIRNVLASASADNTVILWDMSLGKPAASLAVHTDKVQTLQFHPFEAQTLISGSYDKSVALYDCRSPDESHRMWRFSGQIERVTWNHFSPCHFLASTDDGFVYNLDARSDKPIFTLNAHNDEISGLDLSSQIKGCLVTASADKYVKIWDILGDRPSLVHSRDMKMGVLFCSSCCPDLPFIYALRGQKEGLRVWDISTVSSVNEAFGRRERLVLGSARNSSISGPFGSRSSDTPMESSpecifications
PWP1 homolog (S. cerevisiae) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PWP1 protein. | |
PWP1 | |
IEF-SSP-9502 | |
Mouse | |
50 μL | |
Yes | |
11137 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence,Western Blot | |
Unconjugated | |
No additive | |
BC001652.2 | |
PWP1 | |
PWP1 (AAH01652.1, 1 a.a. ∼ 501 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |