Please login to your online account to display your discounted pricing

anti-PUMA, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081760SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081760T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_001120712
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Test Specificity This is specific for isoform 1 of PUMA.
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Gene ID (Entrez) 27113
Antigen PUMA
Host Species Rabbit
Concentration 1mg/mL
Conjugate Unlabeled
Format Affinity Purified
Immunogen Synthetic peptide made to an N-terminal portion of human PUMA (within residues 1-100). [NCBI Sequence NP_001120712]
Quantity 0.025mL