Please login to your online account to display your discounted pricing

anti-PUMA, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081760

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081760

  • / Each
 

Description & Specifications

Specifications

Antigen PUMA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. NP_001120712
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human PUMA (within residues 1-100). [NCBI Sequence NP_001120712]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 27113
Test Specificity This is specific for isoform 1 of PUMA.
Target Species Human
Target Species Murine