Please login to your online account to display your discounted pricing

anti-PUMA, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProt No. Q9BXH1]
Antigen PUMA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500261

 
novus biologicals canada
NB500261
Unlabeled 0.1mL Each for $309.05

NB500261SS

 
novus biologicals canada
NB500261SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProt No. Q9BXH1]
Antigen PUMA
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q9BXH1
Gene ID (Entrez) 27113
Formulation Tris-glycine, 150mM NaCl
Concentration 5.65mg/mL