Please login to your online account to display your discounted pricing

anti-PTPN13/PTPL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056139SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056139T

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation Neat serum
Test Specificity FAP-1 typically migrates at ~270 kDa on western blots. Thus for SDS-PAGE and western blot, the researcher should use techniques optimized for the transfer of proteins with high molecular weights. The actual molecular weight of FAP-1 observed depends on the isoform expression.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5783
Conjugate Unlabeled
Immunogen A recombinant protein corresponding to amino acids 1279 to 1883 of human FAP-1 protein was used as immunogen; GenBank no. NP_542414.1.
Quantity 0.025mL
Cross Reactivity Human
Format Antiserum
Antigen PTPN13/PTPL1