Please login to your online account to display your discounted pricing

anti-PTPN13/PTPL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056139

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056139

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity FAP-1 typically migrates at ~270 kDa on western blots. Thus for SDS-PAGE and western blot, the researcher should use techniques optimized for the transfer of proteins with high molecular weights. The actual molecular weight of FAP-1 observed depends on the isoform expression.
Formulation Neat serum
Gene ID (Entrez) 5783
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.05mL
Primary or Secondary Primary
Immunogen A recombinant protein corresponding to amino acids 1279 to 1883 of human FAP-1 protein was used as immunogen; GenBank no. NP_542414.1.
Cross Reactivity Human
Host Species Rabbit
Antigen PTPN13/PTPL1
Format Antiserum
Regulatory Status RUO