Please login to your online account to display your discounted pricing

anti-PTK7/CCK4, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 900-950 of human PTK7 protein was used as the immunogen.
Antigen PTK7/CCK4
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223655

 
novus biologicals canada
NBP223655
Unlabeled 0.1mL Each for $255.80

NBP223655SS

 
novus biologicals canada
NBP223655SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 900-950 of human PTK7 protein was used as the immunogen.
Antigen PTK7/CCK4
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms A, B, C and D).
Gene Accession No. Q13308
Gene ID (Entrez) 5754
Formulation 100μL of crude rabbit sera with 50% glycerol