Please login to your online account to display your discounted pricing

anti-PTK7/CCK4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223655SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223655SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms A, B, C and D).
Gene Accession No. Q13308
Conjugate Unlabeled
Formulation 100μL of crude rabbit sera with 50% glycerol
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acid 900-950 of human PTK7 protein was used as the immunogen.
Gene ID (Entrez) 5754
Quantity 0.025mL
Format Antiserum
Regulatory Status RUO
Antigen PTK7/CCK4