Please login to your online account to display your discounted pricing

anti-Protocadherin-15, Polyclonal, Novus Biologicals™

$473.61 - $473.61

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the N Terminus Region of the target protein (NM_023115).
Antigen Protocadherin-15
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

2095000201

 
novus biologicals canada
2095000201MG
Unlabeled 0.1mg Each for $473.61
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the N Terminus Region of the target protein (NM_023115).
Antigen Protocadherin-15
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity This product is specific for Human PCDH15.
Gene Accession No. Q96QU1
Gene ID (Entrez) 65217
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0