Please login to your online account to display your discounted pricing

anti-Prothrombin Fragments, Polyclonal, Invitrogen™


Manufacturer: pierce biotechnology inc  PA174090

Catalog No. PIPA174090

  • / Each
 

Description & Specifications

Specifications

Immunogen Prothrombin from human plasma.
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Format Purified
Isotype IgG
Conjugate Unlabeled
Host Species Sheep
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 500μg
Antigen Prothrombin Fragments