Please login to your online account to display your discounted pricing

anti-Protein Phosphatase 2 B alpha Polyclonal, Thermo Scientific™

Protein Phosphatase 2 B alpha Polyclonal Ab PA1-12394 from Thermo Scientific™ detects Protein Phosphatase 2 B alpha in Bv, Hu, Ms and Rt samples, and is validated for IHC and WB

$448.05 - $448.05

Specifications

Antigen Protein Phosphatase 2 B alpha
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-127

 
pierce biotechnology inc
PA112394
Unlabeled 100μg Each for $448.05

01-674-137

 
pierce biotechnology inc
PA112404
Unlabeled 100μg Each for $448.05
Description & Specifications

Specifications

Antigen Protein Phosphatase 2 B alpha
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Sheep
Immunogen Synthetic peptide.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Protein Phosphatase 2 B alpha Polyclonal Ab PA1-12394 from Thermo Scientific™ detects Protein Phosphatase 2 B alpha in Bv, Hu, Ms and Rt samples, and is validated for IHC and WB

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
Host Species: Rabbit
Cross Reactivity: Bovine, Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues R(482) M P P R R D A M P S D A(494) of protein phosphatase 2B alpha.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: PP2CB
Uniprot Accession: P48452, P63328, P63329, Q08209
Gene Symbol: PPP3CA
Entrez GeneID: 19055, 24674, 286852, 5530
PA1-12394 detects Protein Phosphatase 2 B alpha in Bovine, Human, Mouse and Rat samples. PA1-12394 has been successfully used in Immunohistochemistry and Western Blot procedures. PA1-12394 The immunogen is Synthetic peptide corresponding to residues R(482) M P P R R D A M P S D A(494) of protein phosphatase 2B alpha..