Please login to your online account to display your discounted pricing

anti-Protein Phosphatase 2 A, B Polyclonal, Thermo Scientific™

Protein Phosphatase 2 A, B Polyclonal Ab PA1-12389 from Thermo Scientific™ specifically detects Protein Phosphatase 2 A, B in Hu, Ms and Rt samples, and it is validated for WB

$448.05 - $448.05

Specifications

Antigen Protein Phosphatase 2 A-B alpha
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-120

 
pierce biotechnology inc
PA112387
Unlabeled 100μg Each for $448.05

01-674-122

 
pierce biotechnology inc
PA112389
Unlabeled 100μg Each for $448.05

01-674-119

 
pierce biotechnology inc
PA112386
Unlabeled 100μg Each for $448.05
Description & Specifications

Specifications

Antigen Protein Phosphatase 2 A-B alpha
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Sheep
Immunogen Synthetic peptide.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Protein Phosphatase 2 A, B Polyclonal Ab PA1-12389 from Thermo Scientific™ specifically detects Protein Phosphatase 2 A, B in Hu, Ms and Rt samples, and it is validated for WB

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide, conjugated to KLH, corresponding to the N-terminus of PP2A/B delta.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: PP2CB
Uniprot Accession: Q14738
Gene Symbol: PPP2R5D
Entrez GeneID: 21770, 363193, 5528
PA1-12389 detects Protein Phosphatase 2 A, B in Human, Mouse and Rat samples. PA1-12389 has been successfully used in Western Blot procedures. PA1-12389 The immunogen is Synthetic peptide, conjugated to KLH, corresponding to the N-terminus of PP2A/B delta..