missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PPP2R1A Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112384

Catalog No. 01-674-117



Description

Description

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.
Specifications

Specifications

PPP2R1A
Polyclonal
PBS with 0.08% sodium azide
Q32PI5, P30153, Q76MZ3
PP2Ac, PP2CA, PP2Calpha, RP-C
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
117281, 51792, 535321, 5518
Immunohistochemistry, Western Blot
Unconjugated
PPP2R1A
Liquid
PPP2R1A
Synthetic peptide.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
Documents