Please login to your online account to display your discounted pricing

anti-Protein Phosphatase 2 A, A Polyclonal, Thermo Scientific™

Protein Phosphatase 2 A, A Polyclonal Ab PA1-12384 from Thermo Scientific™ detects Protein Phosphatase 2 A, A in Bv, Hu, Ms and Rt samples, and is validated for IHC and WB

$448.05 - $448.05

Specifications

Antigen Protein Phosphatase 2 A, A
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-674-117

 
pierce biotechnology inc
PA112384
Unlabeled 100μg Each for $448.05

01-674-118

 
pierce biotechnology inc
PA112385
Unlabeled 100μg Each for $448.05
Description & Specifications

Specifications

Antigen Protein Phosphatase 2 A, A
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Protein Phosphatase 2 A, A Polyclonal Ab PA1-12384 from Thermo Scientific™ detects Protein Phosphatase 2 A, A in Bv, Hu, Ms and Rt samples, and is validated for IHC and WB

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.
Host Species: Rabbit
Cross Reactivity: Bovine, Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide.
Formulation: 100μg purified Antibody in PBS, with 0.08% sodium azide;
Synonyms: PP2Ac, PP2CA, PP2Calpha, RP-C
Uniprot Accession: P30153, Q32PI5, Q76MZ3
Gene Symbol: PPP2R1A
Entrez GeneID: 117281, 51792, 535321, 5518
PA1-12384 detects Protein Phosphatase 2 A, A in Bovine, Human, Mouse and Rat samples. PA1-12384 has been successfully used in Immunohistochemistry and Western Blot procedures. PA1-12384 The immunogen is Synthetic peptide..