missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Protein Kinase D2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 170-210 of human PKD2 was used as the immunogen.
Antigen Protein Kinase D2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224592 Novus Biologicals Canada
NBP224592
0.1mg Each for $257.41
NBP224592SS Novus Biologicals Canada
NBP224592SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Protein Kinase D2
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, dog, horse, mouse, opossum, rat, 94% homologous in cow and 77% homologous in zebrafish.
100μg in 200μL PBS containing 0.05% BSA
A portion of amino acids 170-210 of human PKD2 was used as the immunogen.
Polyclonal
Primary
Western Blot
25865
0.5mg/mL