Please login to your online account to display your discounted pricing

anti-Protein Kinase D2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224592SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224592SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 25865
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, dog, horse, mouse, opossum, rat, 94% homologous in cow and 77% homologous in zebrafish.
Concentration 0.5mg/mL
Host Species Rabbit
Immunogen A portion of amino acids 170-210 of human PKD2 was used as the immunogen.
Applications Western Blotting
Antigen Protein Kinase D2
Quantity 0.025mg
Regulatory Status RUO