Please login to your online account to display your discounted pricing

anti-Profilin 1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of human Profilin 1 (within residues 100-140). [Swiss-Prot No. P07737]
Antigen Profilin 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP119344

 
novus biologicals canada
NBP119344
Unlabeled 0.1mL Each for $309.05

NBP119344SS

 
novus biologicals canada
NBP119344SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of human Profilin 1 (within residues 100-140). [Swiss-Prot No. P07737]
Antigen Profilin 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Gene Accession No. P07737
Gene ID (Entrez) 5216
Formulation PBS, 30% glycerol
Concentration 1.37mg/mL