Please login to your online account to display your discounted pricing

anti-Profilin 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP119344SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP119344SS

  • / Each
 

Description & Specifications

Specifications

Antigen Profilin 1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 1.37mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P07737
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of human Profilin 1 (within residues 100-140). [Swiss-Prot No. P07737]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5216
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Rat