Please login to your online account to display your discounted pricing

anti-PRMT7, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Amino acids 346-360 of human PRMT7 were used as the immunogen.
Antigen PRMT7
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056400

 
novus biologicals canada
NB10056400
Unlabeled 0.1mg Each for $307.00

NB10056400T

 
novus biologicals canada
NB10056400SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 346-360 of human PRMT7 were used as the immunogen.
Antigen PRMT7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen can be found in five different human PRMT7 isoforms: CRA_a (692 amino acids), CRA_b (613 amino acids), CRA_c (465 amino acids), CRA_d (655 amino acids) and CRA_e (567 amino acids).
Gene Accession No. Q9NVM4
Gene ID (Entrez) 54496
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL