Please login to your online account to display your discounted pricing

anti-PRMT6, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed a synthetic peptide corresponding to amino acids 23-43 (CEEDGAEREAALERPRRTKRER) of human PRMT6.
Antigen PRMT6
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056642

 
novus biologicals canada
NB10056642
Unlabeled 0.2mL Each for $329.00

NB10056642T

 
novus biologicals canada
NB10056642SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed a synthetic peptide corresponding to amino acids 23-43 (CEEDGAEREAALERPRRTKRER) of human PRMT6.
Antigen PRMT6
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. Q96LA8
Gene ID (Entrez) 55170
Formulation PBS containing 0.05% BSA