Please login to your online account to display your discounted pricing

anti-PRMT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056642SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056642T

  • / Each
 

Description & Specifications

Specifications

Antigen PRMT6
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Antisera
Formulation PBS containing 0.05% BSA
Gene Accession No. Q96LA8
Host Species Rabbit
Immunogen This antibody was developed a synthetic peptide corresponding to amino acids 23-43 (CEEDGAEREAALERPRRTKRER) of human PRMT6.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55170
Target Species Human
Target Species Murine