Please login to your online account to display your discounted pricing

anti-PRMT2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11093593

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11093593

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity This is specific for PRMT2, and may also recognize a homodimer at ∽98 kDa as referenced in Chao et al (3). Human and orangutan. Immunogen sequence has 94% homology to bovine.
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Gene Accession No. P55345
Gene ID (Entrez) 3275
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Primate
Antigen PRMT2
Immunogen Synthetic peptide made to an internal portion of human PRMT2 (within residues 380-433). [Swiss-Prot No. P55345]