Please login to your online account to display your discounted pricing

anti-PRMT2, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human PRMT2 (within residues 380-433). [Swiss-Prot No. P55345]
Antigen PRMT2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11093593

 
novus biologicals canada
NB11093593
Unlabeled 0.1mL Each for $307.00

NB11093593T

 
novus biologicals canada
NB11093593SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human PRMT2 (within residues 380-433). [Swiss-Prot No. P55345]
Antigen PRMT2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity This is specific for PRMT2, and may also recognize a homodimer at ∽98 kDa as referenced in Chao et al (3). Human and orangutan. Immunogen sequence has 94% homology to bovine.
Gene Accession No. P55345
Gene ID (Entrez) 3275
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL