Please login to your online account to display your discounted pricing

anti-PRMT2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11093593SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11093593T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P55345
Quantity 0.025mL
Gene ID (Entrez) 3275
Test Specificity This is specific for PRMT2, and may also recognize a homodimer at ∽98 kDa as referenced in Chao et al (3). Human and orangutan. Immunogen sequence has 94% homology to bovine.
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Primate
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Immunogen Synthetic peptide made to an internal portion of human PRMT2 (within residues 380-433). [Swiss-Prot No. P55345]
Antigen PRMT2
Format Affinity Purified
Host Species Rabbit
Regulatory Status RUO