Please login to your online account to display your discounted pricing

anti-Prion protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056606

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056606

  • / Each
 

Description & Specifications

Specifications

Antigen Prion protein
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Sheep
Format Purified
Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen Amino acids 217-229 of BSE protein were used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5621
Test Specificity The NCBI accession number for BSE protein is AAQ64648.
Target Species Bovine
Target Species Sheep