Please login to your online account to display your discounted pricing

anti-Prion protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056605

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056605

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Cross Reactivity Avian
Cross Reactivity Bovine
Cross Reactivity Sheep
Immunogen Amino acids 162-170 of BSE protein were used as the immunogen.
Gene ID (Entrez) 5621
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Applications Western Blotting
Antigen Prion protein
Quantity 0.1mg
Format Purified
Host Species Rabbit
Regulatory Status RUO