Please login to your online account to display your discounted pricing

anti-Prion protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056604SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056604T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 5621
Host Species Rabbit
Regulatory Status RUO
Format Purified
Formulation PBS containing 0.2% gelatin
Quantity 0.025mg
Cross Reactivity Bovine
Cross Reactivity Sheep
Antigen Prion protein
Immunogen Amino acids 142-148 of BSE protein were used as the immunogen.