missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Prion protein, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Amino acids 142-148 of BSE protein were used as the immunogen.
Antigen Prion protein
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056604 Novus Biologicals Canada
NB10056604
0.1mg Each for $259.02
NB10056604T Novus Biologicals Canada
NB10056604SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
Prion protein
Purified
RUO
5621
0.5mg/mL
Amino acids 142-148 of BSE protein were used as the immunogen.
Polyclonal
Primary
Western Blot
PBS containing 0.2% gelatin