Please login to your online account to display your discounted pricing

anti-PRDM5, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human PRDM5 protein were used as the immunogen.
Antigen PRDM5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056419

 
novus biologicals canada
NB10056419
Unlabeled 0.1mg Each for $255.80

NB10056419T

 
novus biologicals canada
NB10056419SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human PRDM5 protein were used as the immunogen.
Antigen PRDM5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Cross reacts with Human. Predicted to cross react with Canine.
Gene Accession No. Q9NQX1
Gene ID (Entrez) 11107
Formulation PBS containing 0.2% gelatin