Please login to your online account to display your discounted pricing

anti-PRAME, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP103359

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP103359

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 23532
Gene Accession No. P78395
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Antigen PRAME
Formulation Tris-citrate/phosphate, pH 7-8
Immunogen Synthetic peptide made to an N-terminal portion of human PRAME (within residues 1-100). [Swiss-Prot No. P78395]
Regulatory Status RUO
Cross Reactivity Human
Format Affinity Purified
Host Species Rabbit
Quantity 0.1mL
Applications Western Blotting