Please login to your online account to display your discounted pricing

anti-PRAME, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human PRAME (within residues 1-100). [Swiss-Prot No. P78395]
Antigen PRAME
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP103359

 
novus biologicals canada
NBP103359
Unlabeled 0.1mL Each for $255.80

NBP103359SS

 
novus biologicals canada
NBP103359SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human PRAME (within residues 1-100). [Swiss-Prot No. P78395]
Antigen PRAME
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. P78395
Gene ID (Entrez) 23532
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL