Please login to your online account to display your discounted pricing

anti-PPAR gamma/NR1C3, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PPAR gamma protein (between residues 20-120) [UniProt P37231]
Antigen PPAR gamma/NR1C3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222106

 
novus biologicals canada
NBP222106
Unlabeled 0.1mL Each for $329.00

NBP222106SS

 
novus biologicals canada
NBP222106SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PPAR gamma protein (between residues 20-120) [UniProt P37231]
Antigen PPAR gamma/NR1C3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Immunogen has 87% homology to rat.
Gene Accession No. P37231
Gene ID (Entrez) 5468
Formulation PBS
Concentration 1.0mg/mL