Please login to your online account to display your discounted pricing

anti-PPAR gamma/NR1C3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222106SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222106SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Concentration 1.0mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Cross Reactivity Human
Cross Reactivity Mouse
Antigen PPAR gamma/NR1C3
Gene ID (Entrez) 5468
Regulatory Status RUO
Format Affinity Purified
Gene Accession No. P37231
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Immunogen has 87% homology to rat.
Immunogen A synthetic peptide made to an internal portion of the human PPAR gamma protein (between residues 20-120) [UniProt P37231]
Quantity 0.025mL
Host Species Rabbit