Please login to your online account to display your discounted pricing

anti-PPAR gamma/NR1C3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222106

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222106

  • / Each
 

Description & Specifications

Specifications

Test Specificity Immunogen has 87% homology to rat.
Formulation PBS
Gene Accession No. P37231
Gene ID (Entrez) 5468
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Concentration 1.0mg/mL
Conjugate Unlabeled
Antigen PPAR gamma/NR1C3
Host Species Rabbit
Regulatory Status RUO
Format Affinity Purified
Immunogen A synthetic peptide made to an internal portion of the human PPAR gamma protein (between residues 20-120) [UniProt P37231]
Cross Reactivity Human
Cross Reactivity Mouse