Please login to your online account to display your discounted pricing

anti-Potassium Channel Kv3.1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142819

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142819

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Conjugate Unlabeled
Test Specificity Does not cross-react with Kv1.2 or Kv1.3. Human, mouse, rat, rabbit, bovine, canine and other vertebrates.
Gene ID (Entrez) 3746
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of rat Kv3.1 (within residues 350-400). [Swiss-Prot No. P25122]
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Applications Blocking
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Format Affinity Purified
Primary or Secondary Primary
Concentration 1mg/mL
Regulatory Status RUO
Quantity 0.1mL
Antigen Potassium Channel Kv3.1