Please login to your online account to display your discounted pricing

anti-POT1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056429

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056429

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Applications Gel Supershift
Applications Western Blotting
Test Specificity The amino acid sequence used is 100% homologous in human, chimp and monkey, and 86% in bovine and dog. This antibody will detect variant 1 (634 amino acids) and variant 3 (518 amino acids) of POT1. Reacts with Human. Predicted to react with Bovine, Chimpanzee, Canine and New World Monkey.
Concentration 0.5mg/mL
Antigen POT1
Host Species Rabbit
Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Gene Accession No. AAW22613.1
Quantity 0.1mg
Gene ID (Entrez) 25913
Format Purified
Immunogen A portion of amino acids 450-500 of human POT1 was used as the immunogen.
Regulatory Status RUO
Cross Reactivity Human