missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-POT1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human POT1 was used as the immunogen.
Antigen POT1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056429 Novus Biologicals Canada
NB10056429
0.1mg Each for $255.80
NB10056429T Novus Biologicals Canada
NB10056429SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
POT1
Purified
RUO
The amino acid sequence used is 100% homologous in human, chimp and monkey, and 86% in bovine and dog. This antibody will detect variant 1 (634 amino acids) and variant 3 (518 amino acids) of POT1. Reacts with Human. Predicted to react with Bovine, Chimpanzee, Canine and New World Monkey.
25913
0.5mg/mL
A portion of amino acids 450-500 of human POT1 was used as the immunogen.
Polyclonal
Primary
Gel Supershift,Western Blot
AAW22613.1
PBS containing 0.05% BSA